Learn More
Iodonitrotetrazolium Chloride, Sigma-Aldrich
Catalog No. 01259031 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-031 5 g
01-258-976 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-031 Supplier MilliporeSigma Supelco Supplier No. I83775G
Add to Cart
Add to Cart
Histochemical reagent used as an electron acceptor in colorimetric assays for dehydrogenases.
Electron acceptor for the colorimetric assay of various dehydrogenases
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 g
Product Type Stain Kit