Learn More
Crescent Chemical Co Inc IODONITROTETRAZOLIUM CHLORIDE
SDP
Catalog No. 50247760
Encompass_Preferred
Iodonitrotetrazolium chloride research grade For LDH detection (1).For colorimetric measurement of enzymatic hydrolysis of terminal galactose from GM1 ganglioside (2). Size - 2.5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 26840.02 CAS 146-
Catalog No. 50-247-760 Supplier Crescent Chemical Co Inc Supplier No. SE26840.02
Iodonitrotetrazolium chloride research grade For LDH detection (1).For colorimetric measurement of enzymatic hydrolysis of terminal galactose from GM1 ganglioside (2). Size - 2.5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 26840.02 CAS 146-