Learn More
Cayman Chemical Iodonitrotetrazolium (chloride), Purity: ≥98%, CAS: 146-68-9, 1 g
SDP
Catalog No. 502992657
Encompass_Preferred
A monotetrazolium salt used as an indicator dye; reduced to an insoluble formazan that is used as a vital dye or indicator of cellular redox activity; commonly used to measure the respiratory activity of microorganisms in a variety of contexts
This item is not returnable. View return policy
Catalog No. 50-299-2657 Supplier Cayman Chemical Supplier No. 160731 G
A monotetrazolium salt used as an indicator dye; reduced to an insoluble formazan that is used as a vital dye or indicator of cellular redox activity; commonly used to measure the respiratory activity of microorganisms in a variety of contexts