Learn More
Medchemexpress LLC Iodopropynyl butylcarbamate | 55406-53-6 | 99.48% | 281.09 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW009335100G
Encompass_Preferred
Iodopropynyl butylcarbamate (IPBC) is a potent fungicide and preservative used in various industries. It is primarily used as a laboratory chemical and for the manufacture of substances.
  • Potent fungicide and preservative
  • Used in paints, coatings, and wood preservatives
  • Used in personal care and cosmetics industries
Catalog No. 50-003-66953