Learn More
Medchemexpress LLC Iodopropynyl butylcarbamate | 55406-53-6 | 99.5% | 281.09 | 10 G
SDP
Catalog No. 5000452636
Encompass_Preferred
Iodopropynyl butylcarbamate (IPBC) is a potent fungicide and preservative used across various industries, including paints, coatings, wood preservatives, personal care, and cosmetics. This chemical compound is presented as a solid with a white to off-white appearance.
  • Functions as a potent fungicide and preservative.
  • Suitable for paints, coatings, and wood preservation.
  • Applicable in personal care and cosmetics industries.
  • Appears as a white to off-white solid.
  • High purity for reliable applications.
Catalog No. 50-004-52636 Supplier Medchemexpress LLC Supplier No. HYW00933510G
Add to Cart
Add to Cart
Iodopropynyl butylcarbamate (IPBC) is a potent fungicide and preservative used across various industries, including paints, coatings, wood preservatives, personal care, and cosmetics. This chemical compound is presented as a solid with a white to off-white appearance.
  • Functions as a potent fungicide and preservative.
  • Suitable for paints, coatings, and wood preservation.
  • Applicable in personal care and cosmetics industries.
  • Appears as a white to off-white solid.
  • High purity for reliable applications.