Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoquinol, Powder, USP, 96-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18603495 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-495 25 g Amber Glass Bottle
18-603-496 100 g Amber Glass Bottle
18-603-499 500 g Amber Glass Bottle
18-603-497 1 kg Amber Glass Bottle
18-603-498 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-495 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. IO11525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

IO115, 83-73-8, C9H5I2NO

Spectrum™ Chemical Iodoquinol, Powder, USP Also known as Diquinol and iodoxin, is used in the treatment of amoebiasis and as a luminal amebicide. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 83-73-8
Molecular Formula C9H5I2NO
Molecular Weight (g/mol) 396.95
InChI Key UXZFQZANDVDGMM-UHFFFAOYSA-N
IUPAC Name 5,7-diiodoquinolin-8-ol
SMILES OC1=C2N=CC=CC2=C(I)C=C1I

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 96 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.