Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodosobenzene diacetate, 98+%

Catalog No. aab2453114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2453114 25 g
AAB2453122 100 g
AAB2453136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2453114 Supplier Thermo Scientific Chemicals Supplier No. B2453114
Only null left
Add to Cart

CAS: 3240-34-4 | C10H11IO4 | 322.098 g/mol

Iodosobenzene is used as an oxidizing and acetoxylating agent in organic synthesis. It is actively involved in the preparation of (Z)-3,7-dimethyl-2,6-octadien-1-al(neral) from (Z)-3,7-dimethyl-2,6-octadien-1-ol (nerol) in presence of 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO). It is a useful reagent for the synthesis of a wide variety of heterocyclic compounds. It is also used in the Pd-catalyzed 2-arylation of indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodosobenzene is used as an oxidizing and acetoxylating agent in organic synthesis. It is actively involved in the preparation of (Z)-3,7-dimethyl-2,6-octadien-1-al(neral) from (Z)-3,7-dimethyl-2,6-octadien-1-ol (nerol) in presence of 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO). It is a useful reagent for the synthesis of a wide variety of heterocyclic compounds. It is also used in the Pd-catalyzed 2-arylation of indoles.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3240-34-4
Molecular Formula C10H11IO4
Molecular Weight (g/mol) 322.098
MDL Number MFCD00008692
InChI Key ZBIKORITPGTTGI-UHFFFAOYSA-N
Synonym iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo
PubChem CID 76724
IUPAC Name [acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
SMILES CC(=O)OI(C1=CC=CC=C1)OC(=O)C

Specifications

Melting Point 159°C to 161°C
Quantity 25 g
Beilstein 1879369
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 322.1
Percent Purity ≥98%
Chemical Name or Material Iodosobenzene diacetate
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 221-808-1
RTECSNumber DA3525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.