Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodosodilactone 98.0+%, TCI America™
SDP

Catalog No. I08651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08651G Supplier TCI America Supplier No. I08651G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2902-68-3
Molecular Formula C8H3IO4
Molecular Weight (g/mol) 290.012
InChI Key DHIWEENQJCGNME-UHFFFAOYSA-N
Synonym 2H,6H-[1,2]Iodoxolo[4,5,1-hi]benziodoxole-2,6-dione
PubChem CID 60153134
SMILES C1=CC2=C3C(=C1)C(=O)OI3OC2=O

Specifications

Melting Point 263°C
Color White-Yellow
Quantity 1 g
Formula Weight 290.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Iodosodilactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.