Learn More
Med Vet International Iodosorb Antimicrobial Wound Gel, 40-gram Tube, 6602125040, Each
Catalog No. 503044600
Encompass_Preferred
Antimicrobial Wound Gel Iodosorb Dual-action design helps create an effective wound healing environment Offers the benefits of a broad-spectrum slow-release antimicrobial agent in combination with desloughing and fluid management properties Prepares the wound bed to heal in highly exuding chronic wounds and infected wounds Removes loose slough and debris to help clean the wound bed 1.4-ounce tube
Catalog No. 50-304-4600 Supplier Med Vet International Supplier No. 549315EA
Antimicrobial Wound Gel Iodosorb Dual-action design helps create an effective wound healing environment Offers the benefits of a broad-spectrum slow-release antimicrobial agent in combination with desloughing and fluid management properties Prepares the wound bed to heal in highly exuding chronic wounds and infected wounds Removes loose slough and debris to help clean the wound bed 1.4-ounce tube