Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iodotrimethylsilane Solution

Catalog No. 748854100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-885-4100ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 74-885-4100ML Supplier Sigma Aldrich Supplier No. 748854100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iodotrimethylsilane [(CH3)3SiI] is an important and versatile organosilicon reagent with wide applications in organic synthesis. It is used as a neutral nucleophilic reagent, trimethylsilylating agent, Lewis acid catalyst, reducing agent and dehydrating agent in many organic reactions.Iodotrimethylsilane can be used as:A reagent in the hydroiodation of ynamides to yield the important building blocks (E)-+a-iodoenamides.A reagent in the synthesis of (1-iodovinyl) arenes by hydroiodation of trimethylsilyl ethynylarenes.

Specifications

CAS 5464-68-6
Molecular Formula C6H14NO4P
Linear Formula C6H14NO4P
MDL Number MFCD00014805
Quantity 1 g
Synonym Iodotrimethylsilane solution; TMS iodine; TMS-I; TMSI; Trimethyliodosilane
Molecular Weight (g/mol) 195.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.