Learn More
Abcam Iodotubercidin, adenosine kinase inhibitor, 1MG
Catalog No. 502707585
Encompass_Preferred
MW 392.15 Da, Purity >95%. Potent, specific adenosine kinase inhibitor (IC₅₀ = 26 nM). Inhibits platelet aggregation and spreading, and interferes with adenosine and adenine nucleotide metabolism. Additionally inhibits other kinases such as ERK2 (Ki = 525 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7585 Supplier Abcam Supplier No. AB1204521MG
MW 392.15 Da, Purity >95%. Potent, specific adenosine kinase inhibitor (IC₅₀ = 26 nM). Inhibits platelet aggregation and spreading, and interferes with adenosine and adenine nucleotide metabolism. Additionally inhibits other kinases such as ERK2 (Ki = 525 nM).
The product is subject to the following: Abcam Restricted Use Statement