Learn More
Medchemexpress LLC HY-B0594 100mg Medchemexpress, Iohexol CAS:66108-95-0 Purity:>98%
SDP
Catalog No. 502082615
Encompass_Preferred
Medchemexpress, HY-B0594 100mg Iohexol CAS:66108-95-0 Iohexol is a radiographic contrast agent and can be applied for myelography, computerized tomography (cisternography, ventriculography) and MicroCT imaging in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2615 Supplier Medchemexpress LLC Supplier No. HYB0594100MG
Medchemexpress, HY-B0594 100mg Iohexol CAS:66108-95-0 Iohexol is a radiographic contrast agent and can be applied for myelography, computerized tomography (cisternography, ventriculography) and MicroCT imaging in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.