Learn More
Medchemexpress LLC Iohexol | 66108-95-0 | 99.2% | 100 MG
SDP
Catalog No. 5000405896
Encompass_Preferred
Iohexol (Standard) is the analytical standard of Iohexol. This product is intended for research and analytical applications. Iohexol is a non-ionic, water-soluble contrast agent used as a reference marker for the determination of glomerular filtration rate (GFR) at the level of renal function. Iohexol can be used for contrast in myelography, computerized tomography (cisternography, ventriculography) and MicroCT imaging.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-05896 Supplier Medchemexpress LLC Supplier No. HYB0594R100MG
Iohexol (Standard) is the analytical standard of Iohexol. This product is intended for research and analytical applications. Iohexol is a non-ionic, water-soluble contrast agent used as a reference marker for the determination of glomerular filtration rate (GFR) at the level of renal function. Iohexol can be used for contrast in myelography, computerized tomography (cisternography, ventriculography) and MicroCT imaging.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.