Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iohexol | 66108-95-0 | MFCD00077732 | 99.2% | 821.14 g/mol | C19H26I3N3O9 | 10 MG
SDP

Catalog No. 5000298299
Encompass_Preferred

Iohexol analytical standard for laboratory and analytical use. This non-ionic, water-soluble contrast agent is supplied as an analytical reference standard for method development, calibration, and quantitative assays (HPLC, GC, MS). The material is intended for research and analytical applications only and is provided in small milligram pack sizes suitable for laboratory workflows.

  • Analytical reference standard for iohexol.
  • High purity suitable for quantitative analysis.
  • Molecular formula C19H26I3N3O9; molecular weight 821.14 g/mol.
  • Available in small milligram pack sizes for method development and calibration.
  • Intended for research and analytical use; not for diagnostic or clinical administration.

Catalog No. 50-002-98299 Supplier Medchemexpress LLC Supplier No. HYB0594R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iohexol analytical standard for laboratory and analytical use. This non-ionic, water-soluble contrast agent is supplied as an analytical reference standard for method development, calibration, and quantitative assays (HPLC, GC, MS). The material is intended for research and analytical applications only and is provided in small milligram pack sizes suitable for laboratory workflows.

  • Analytical reference standard for iohexol.
  • High purity suitable for quantitative analysis.
  • Molecular formula C19H26I3N3O9; molecular weight 821.14 g/mol.
  • Available in small milligram pack sizes for method development and calibration.
  • Intended for research and analytical use; not for diagnostic or clinical administration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.