Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iohexol, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015106 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5106 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5106 Supplier MilliporeSigma Supelco Supplier No. 7414750MG
Only null left
Add to Cart
Add to Cart

Iohexol is a non-ionic, water-soluble contrast agent used as a reference marker to determine the glomerular filtration rate (GFR) which measures the level of kidney function. It finds applications in angiographic and urography procedures.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 66108-95-0
Physical Form Neat
Linear Formula C19H26I3N3O9
MDL Number MFCD00077732
Synonym 5-[N-(2,3-Dihydroxypropyl)acetamido]-2,4,6-triiodo-N,N′-bis(2,3-dihydroxypropyl)isophthalamide
Type Analytical Standard
Quantity 50 mg
Formula Weight 821.14
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.