Learn More
Medchemexpress LLC Iohexol-d5 | 928623-33-0 | 100.0% | 1 MG
SDP
Catalog No. 5000406881
Encompass_Preferred
Iohexol-d5 is a deuterium-labeled derivative of Iohexol, a widely recognized radiographic contrast agent. This product is suitable for various imaging applications including myelography, computerized tomography, and MicroCT imaging. It also serves as a valuable tool in quantitative analysis as a tracer or internal standard for techniques like NMR, GC-MS, and LC-MS.
  • Deuterium-labeled compound for research applications
  • Functions as a radiographic contrast agent
  • Useful as a tracer in analytical studies
  • Can be used as an internal standard for quantitative analysis
  • Suitable for myelography and computerized tomography
Catalog No. 50-004-06881 Supplier Medchemexpress LLC Supplier No. HYB0594S1MG
Iohexol-d5 is a deuterium-labeled derivative of Iohexol, a widely recognized radiographic contrast agent. This product is suitable for various imaging applications including myelography, computerized tomography, and MicroCT imaging. It also serves as a valuable tool in quantitative analysis as a tracer or internal standard for techniques like NMR, GC-MS, and LC-MS.
  • Deuterium-labeled compound for research applications
  • Functions as a radiographic contrast agent
  • Useful as a tracer in analytical studies
  • Can be used as an internal standard for quantitative analysis
  • Suitable for myelography and computerized tomography