Learn More
Medchemexpress LLC Iohexol (Standard) | 66108-95-0 | 99.2% | 50 MG
SDP
Catalog No. 5000408331
Encompass_Preferred
Iohexol (Standard) is the analytical standard for Iohexol, intended for research and analytical applications. It functions as a non-ionic, water-soluble contrast agent. Iohexol is utilized as a reference marker to determine the glomerular filtration rate (GFR) at the renal function level. Additionally, it can be employed for contrast in various imaging procedures including myelography, computerized tomography (cisternography, ventriculography), and MicroCT imaging.
  • Analytical standard for Iohexol
  • Non-ionic
  • Water-soluble contrast agent
  • Used as a reference marker for glomerular filtration rate (GFR)
  • Applicable in myelography, computerized tomography (cisternography, ventriculography), and MicroCT imaging
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-08331 Supplier Medchemexpress LLC Supplier No. HYB0594R50MG
Iohexol (Standard) is the analytical standard for Iohexol, intended for research and analytical applications. It functions as a non-ionic, water-soluble contrast agent. Iohexol is utilized as a reference marker to determine the glomerular filtration rate (GFR) at the renal function level. Additionally, it can be employed for contrast in various imaging procedures including myelography, computerized tomography (cisternography, ventriculography), and MicroCT imaging.
  • Analytical standard for Iohexol
  • Non-ionic
  • Water-soluble contrast agent
  • Used as a reference marker for glomerular filtration rate (GFR)
  • Applicable in myelography, computerized tomography (cisternography, ventriculography), and MicroCT imaging
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS