Learn More
Medchemexpress LLC Iohexol (Standard) | 66108-95-0 | MFCD00077732 | 99.54% | 25 MG
SDP
Catalog No. 5000450958
Encompass_Preferred
Iohexol (Standard) is the analytical standard of Iohexol, intended for research and analytical applications. It is a non-ionic, water-soluble contrast agent used as a reference marker for determining glomerular filtration rate (GFR) at the renal function level. Iohexol can be used for contrast in myelography, computerized tomography (cisternography, ventriculography), and MicroCT imaging.
  • Analytical standard of Iohexol
  • Non-ionic, water-soluble contrast agent
  • Used as a reference marker for glomerular filtration rate (GFR)
  • Suitable for contrast in myelography, computerized tomography, and MicroCT imaging
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-50958 Supplier Medchemexpress LLC Supplier No. 25MGHYB0594R
Iohexol (Standard) is the analytical standard of Iohexol, intended for research and analytical applications. It is a non-ionic, water-soluble contrast agent used as a reference marker for determining glomerular filtration rate (GFR) at the renal function level. Iohexol can be used for contrast in myelography, computerized tomography (cisternography, ventriculography), and MicroCT imaging.
  • Analytical standard of Iohexol
  • Non-ionic, water-soluble contrast agent
  • Used as a reference marker for glomerular filtration rate (GFR)
  • Suitable for contrast in myelography, computerized tomography, and MicroCT imaging
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS