Learn More
ACCU-SCOPE™ IOL-100 Portable Microscope
SDP
Catalog No. 01189185 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Objective Types:
Fixed
Catalog No. Objective Types
01-189-185 Fixed
1 options
Catalog No. 01-189-185 Supplier ACCU-SCOPE™ Supplier No. IOL100
ioLight Digital ultra-portable, compact, and rechargeable digital microscopes deliver live HD images directly to a mobile device, ideal for field study and education. With top and bottom illumination and accepting standard glass microscope slides and coverslips.
  • View live images on your smart phone or computer via WiFi connection
  • 1 mm field of view 1 μm resolution Capture 5 MP JPG images
  • Capture video at 10 fps up to 400x magnification (depending on size of mobile device screen)
  • Working distance: 1 mm
  • Variable top and bottom illumination
  • Glass stage, easy to clean
  • Operate microscope with Apple or Android mobile device or via browser.
  • Rechargeable Li-ion battery via USB charger
  • Optional travel case (CAT #IOL-3295)
Specifications
Eyepiece Fixed
Lighting Type LED
Head Style Monocular
Voltage 5 V
Magnification Power 400X
Objective Types Fixed