Learn More
ATCC IOMM-Lee; Meningioma; Human (Homo sapiens)
Supplier:  ATCC CRL3370
Encompass
IOMM-Lee is a cell line with epithelial-like morphology that was isolated from the tentorial brain of a 61-year-old male with meningioma. It has applications for neuroscience research.
Catalog No. 50-238-2926