Learn More
Ion Torrent™ Ion 520™ Chip Kit-4 Reactions

Description
The Ion 520™ Chip Kit-4 Reactions contains 4 barcoded chips for sample tracking and sequencing of 400-600 bp fragments, for use with the Ion 520™ & Ion 530™ ExT Kit-Chef and the Ion S5™ and Ion S5™ XL Sequencing Systems. The Ion 520 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of as little as 4.5 hours for 600 bp sequencing.
The Ion 520 Chip Kit-4 Reactions offers:
- Sample tracking functionality from an integrated barcode
- A rapid sequencing run time of as little as 4.5 hours, generating 3–4 million reads
- Read lengths of 400-600 bp to support a variety of research applications, including microbial sequencing and metagenomics
- Automated handling and loading of templated products for sequencing
- Compatibility with current library preparation methods
Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL Systems. Use this kit with the Ion 520 & Ion 530 ExT Kit-Chef and Torrent Suite™ Software 5.2.1 or higher.
Specifications
Specifications
| Format | Kit |
| For Use With (Equipment) | Ion GeneStudio S5 System, Ion S5™ XL System |
| For Use With (Application) | Sequencing |
| Number of Reads per Chip | 3-5M |
| Product Line | Ion Torrent |
| Product Type | Chip Kit |
| Quantity | 4 Chips |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.