Learn More
Ion Torrent™ Ion 530™ Chip Kit-4 Reactions

Description
The Ion 530™ Chip Kit-4 Reactions contains 4 barcoded chips for sample tracking and sequencing of 400-600 bp fragments. The kit is intended for use with the Ion 520™ & Ion 530™ ExT Kit-Chef and the Ion S5™ and Ion S5™ XL Sequencing Systems. The Ion 530 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of as little as 4.5 hours for 600 bp sequencing.
The Ion 530 Chip Kit-4 Reactions offers:
- Sample tracking functionality from an integrated barcode
- Read lengths of 400-600 bp to support a variety of research applications, including microbial sequencing and metagenomics
- A rapid sequencing run time of as little as 4.5 hours, generating up to 12 million reads with ExT chemistry
- Automated handling and loading of templated products for sequencing
- Compatibility with current library preparation methods
Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL Systems. Use this kit with the Ion 520 & Ion 530 ExT Kit-Chef and Torrent Suite™ Software 5.2 or higher.
Specifications
Specifications
| Format | Kit |
| For Use With (Equipment) | Ion GeneStudio S5 System, Ion S5™ XL System |
| For Use With (Application) | Sequencing |
| Number of Reads per Chip | 15-20M |
| Product Line | Ion Torrent |
| Product Type | Chip Kit |
| Quantity | 4 Chips |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.