Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Ion AmpliSeq™ Cancer Hotspot Panel v2

Catalog No. 4475346
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-753-46 8 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-753-46 Supplier Ion Torrent™ Supplier No. 4475346
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chip Compatibility Ion 520, Ion 530
Content And Storage Store at -30°C to -10°C.
Sample Type FFPE Tissue, Blood, Cell Cultures, Clinical Samples, DNA (Genomic), DNA from FFPE, dsDNA, Fine Needle Aspirates (FNA), FFPE, Whole Blood
Sequencing Type Next Generation Sequencing
For Use With (Equipment) Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
For Use With (Application) Oncology Research
Libraries Amplicon
Multiplex Capability 207 amplicons
No. of Reactions 8
Nucleic Acid Type DNA
No. of Samples Up to 8 Samples
Product Line Ion AmpliSeq
Product Type Panel
Quantity 8 Reactions
Species Human
Technique Amplicon Sequencing, Post-Light™ Ion Semiconductor Sequencing
Throughput Up to 4 Samples per Ion 510 Chip, Up to 8 Samples per Ion 520 Chip, Up to 26 Samples per Ion 530 Chip
Workflow Step Panel
Starting Material Amount 10 ng DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.