Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Ion AmpliSeq™ Kit for Chef DL8
GSA_VA

Catalog No. A29024
Encompass_Preferred
Change view
Click to view available options
Quantity:
32 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29024 32 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29024 Supplier Ion Torrent™ Supplier No. A29024
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Ion AmpliSeq™ Kit for Chef DL8 is designed for use with the Ion Chef™ System for automation of Ion AmpliSeq library construction.

The Ion AmpliSeq™ Kit for Chef DL8 is designed for use with the Ion Chef™ System for automation of Ion AmpliSeq library construction.

This kit offers:

  • Simplest workflow for targeted sequencing library preparation using 1 or 2 pool Ion AmpliSeq panel designs*
  • Low input
  • Easy-to-use cartridge format with reagent barcoding for full reagent tracking
  • Proven performance of Ion AmpliSeq technology, compatible with DNA extracted from formalin-fixed, paraffin-embedded tissue and other challenging sources

*See AmpliSeq.com for custom 1-2 pool panels. Note that some panels (e.g., Transcriptome (Cat. No. A31446), Cancer Hotspot (A32914), Oncomine BRCA (A32841), Pharmacogenomics (A29998)) have been prepared in an Ion Chef-ready kit by incorporating the respective ready-made primer pool with the DL8 kit.

Leveraging the automation capabilities of the Ion Chef System, the Ion AmpliSeq Kit for Chef DL8 enables the reproducible preparation of Ion AmpliSeq amplicon libraries, with less than 15 minutes of hands-on time and only 3 liquid transfer steps. Compatible with 1- and 2-pool Ion AmpliSeq panel designs, the kit automates the preparation of libraries from 8 samples in a single run, delivering a single tube of pooled libraries ready for downstream templating and sequencing. The kit provides sufficient material for the preparation of 32 libraries (4 Ion Chef runs).

Utilizing plug and play, pre-packaged, single-use cartridges and PCR plates for a fully automated workflow, users of all experience levels will find library preparation straightforward with minimal effort. Supplied with barcodes conveniently dried-down in 96-well plates, multiplexing samples is easy and requires no additional pipetting steps.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Ion AmpliSeq Chef Reagents DL8, 4 cartridges, store at -5°C to -30°C
• Ion AmpliSeq Chef Solutions DL8, 4 cartridges, 2°C to 8°C
• Ion AmpliSeq Supplies (Each box contains: Ion AmpliSeq Tip Cartridge L8, Framed PCR Foil Seal, Enrichment Cartridge), 4 boxes, room temperature
• IonCode in PCR Plates (The set includes: IonCode 0101-0108 in 96-well PCR Plate (red), IonCode 0109-0116 in 96-well PCR Plate (yellow), IonCode 0117-0124 in 96-well PCR Plate (green), IonCode 0125-0132 in 96-well PCR Plate (blue)), 1 set of 4, room temperature
Format Kit
Sample Type DNA
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
For Use With (Application) Sequencing
Libraries Targeted Sequencing Library
No. of Reactions 32 Reactions
Product Line AmpliSeq
Product Type DNA Sequencing Kit
Quantity 32 Reactions
Shipping Condition Dry Ice
Workflow Step Library Generation
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.