Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion AmpliSeq™ Transcriptome Human Gene Expression Kit

Catalog No. A26327
Encompass_Preferred
Change view
Click to view available options
Quantity:
24 Assays
384 Assays
96 Assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26327 384 Assays
A26325 24 Assays
A26326 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A26327 Supplier LIFE TECHNOLOGIES Supplier No. A26327
Add to Cart
Add to Cart
Request a Quote

Specifications

Content And Storage Contains 5X Ion AmpliSeq™ HiFi Mix, FuPa Reagent, Switch Solution.
Store at -5 to -30°C.
Format Kit
Sample Type RNA (Total)
Sequencing Type Transcriptome Sequencing
For Use With (Equipment) Ion Proton™ System
For Use With (Application) Sequencing
Libraries Targeted Sequencing Library
No. of Reactions 384 Assays
Product Line AmpliSeq
Product Type Transcriptome Human Gene Expression Kit
Quantity 384 Assays
Species Human
Workflow Step Library Generation
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.