Learn More
Waters Corp Waters Ion Block Assembly, for ACQUITY QDa Mass Detector, Mechanical, MS System
Catalog No. 502039795 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement ion block assembly for the ACQUITY QDa mass detector. It facilitates the entry of ions into the detector for subsequent detection.
  • Functions as a spare part kit
  • Includes 1 unit per package
  • Features a mechanical type
  • Designed for MS system type
  • Recommended for replacement as required
Catalog No. 50-203-9795 Supplier Waters Corp Supplier No. 700010377
This is a replacement ion block assembly for the ACQUITY QDa mass detector. It facilitates the entry of ions into the detector for subsequent detection.
  • Functions as a spare part kit
  • Includes 1 unit per package
  • Features a mechanical type
  • Designed for MS system type
  • Recommended for replacement as required