Learn More
Waters Corp WATERS CORPORATION Ion Block Assembly, Replacement, for Xevo TQD and SQD2
Catalog No. 50466686 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement ion block assembly designed for use with Xevo TQD and SQD2 systems.
  • Includes sample cone
  • Gas cone
  • Heater cartridge
  • Extractor cone
  • Internal seals
Catalog No. 50-466-686 Supplier Waters Corp Supplier No. 700005999
Add to Cart
Add to Cart
Request a Quote
This is a replacement ion block assembly designed for use with Xevo TQD and SQD2 systems.
  • Includes sample cone
  • Gas cone
  • Heater cartridge
  • Extractor cone
  • Internal seals