Learn More
Waters Corp Waters Ion Block Assembly, Mass Spectrometry, 3100, ACQUITY Compatible
Catalog No. 50792878 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Ion Block Assembly is a replacement mechanical component designed for use in various mass spectrometry systems. It serves as the ion source within the instrument.
  • Compatible with 3100, ACQUITY SQD, ACQUITY TQD, ACQUITY UPLC SQ, and ACQUITY UPLC TQ detectors.
  • Mechanical component for mass spectrometry systems.
  • Supplied as a spare parts kit.
  • Replaced as required for instrument maintenance.
Catalog No. 50-792-878 Supplier Waters Corp Supplier No. 700003523
Add to Cart
Add to Cart
Request a Quote
This Ion Block Assembly is a replacement mechanical component designed for use in various mass spectrometry systems. It serves as the ion source within the instrument.
  • Compatible with 3100, ACQUITY SQD, ACQUITY TQD, ACQUITY UPLC SQ, and ACQUITY UPLC TQ detectors.
  • Mechanical component for mass spectrometry systems.
  • Supplied as a spare parts kit.
  • Replaced as required for instrument maintenance.