Learn More
Waters Corp Waters Corporation Ion Block Assembly, Non-Stepwave, Ion Entry, G2/Qtof/TQ Systems
Catalog No. 50792954 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Ion Block Assembly is designed as a replacement part for Non-Stepwave mass spectrometers. It allows ions to enter the mass spectrometer and is compatible with SYNAPT G2, Xevo G2, Xevo Qtof, and Xevo TQ mass spectrometers.
  • Replacement part for non-stepwave mass spectrometers.
  • Facilitates the entry of ions into the mass spectrometer.
  • Compatible with SYNAPT G2, Xevo G2, Xevo Qtof, and Xevo TQ mass spectrometers.
  • Designed for reliable system performance.
Catalog No. 50-792-954 Supplier Waters Corp Supplier No. 700004203
Add to Cart
Add to Cart
Request a Quote
The Ion Block Assembly is designed as a replacement part for Non-Stepwave mass spectrometers. It allows ions to enter the mass spectrometer and is compatible with SYNAPT G2, Xevo G2, Xevo Qtof, and Xevo TQ mass spectrometers.
  • Replacement part for non-stepwave mass spectrometers.
  • Facilitates the entry of ions into the mass spectrometer.
  • Compatible with SYNAPT G2, Xevo G2, Xevo Qtof, and Xevo TQ mass spectrometers.
  • Designed for reliable system performance.