Learn More
Waters Corp Ion Block Assembly, ETD, SYNAPT G2
Catalog No. 502391388 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Ion Block Assembly, ETD, is a critical replacement component for Non-Stepwave mass spectrometers that are enabled with Electron Transfer Dissociation (ETD) technology. This assembly allows ions to enter the mass spectrometer and includes the necessary ETD needle assembly. It is specifically designed for use with SYNAPT G2 mass spectrometers.
  • Designed to effectively allow ions to enter the mass spectrometer, crucial for accurate analysis.
  • Comes complete with the ETD needle assembly, simplifying replacement and ensuring compatibility.
  • Engineered specifically for SYNAPT G2 mass spectrometers, guaranteeing optimal performance and fit.
  • Supports non-stepwave mass spectrometers enabled with ETD capabilities.
Catalog No. 50-239-1388 Supplier Waters Corp Supplier No. 700005610
Add to Cart
Add to Cart
Request a Quote
The Waters Ion Block Assembly, ETD, is a critical replacement component for Non-Stepwave mass spectrometers that are enabled with Electron Transfer Dissociation (ETD) technology. This assembly allows ions to enter the mass spectrometer and includes the necessary ETD needle assembly. It is specifically designed for use with SYNAPT G2 mass spectrometers.
  • Designed to effectively allow ions to enter the mass spectrometer, crucial for accurate analysis.
  • Comes complete with the ETD needle assembly, simplifying replacement and ensuring compatibility.
  • Engineered specifically for SYNAPT G2 mass spectrometers, guaranteeing optimal performance and fit.
  • Supports non-stepwave mass spectrometers enabled with ETD capabilities.