Learn More
Waters Corp Waters Corporation Ion Block Assembly, Mechanical, MS
Catalog No. 50179761 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement ion block assembly is designed for Stepwave mass spectrometers, facilitating the entry of ions into the mass spectrometer prior to detection.
  • Replacement ion block assembly
  • Designed for Stepwave mass spectrometers
  • Compatible with Xevo TQ-S and SYNAPT G2-S/G2-Si mass spectrometers
  • Mechanical type
  • For mass spectrometry systems
  • Units per package: 1
  • Replacement as required
Catalog No. 50-179-761 Supplier Waters Corp Supplier No. 700005349
Add to Cart
Add to Cart
Request a Quote
This replacement ion block assembly is designed for Stepwave mass spectrometers, facilitating the entry of ions into the mass spectrometer prior to detection.
  • Replacement ion block assembly
  • Designed for Stepwave mass spectrometers
  • Compatible with Xevo TQ-S and SYNAPT G2-S/G2-Si mass spectrometers
  • Mechanical type
  • For mass spectrometry systems
  • Units per package: 1
  • Replacement as required