Learn More
Waters Corp Ion Block Sub Assembly, Stepwave, Mechanical, MS Systems
Catalog No. 502391218 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Ion Block Sub Assembly for Stepwave mass spectrometers is a mechanical component designed to facilitate the entry of ions into the mass spectrometer prior to detection.
  • Mechanical type
  • Designed for MS systems
  • Facilitates ion entry into the mass spectrometer prior to detection
  • Compatible with Xevo TQ-XS, Xevo TQ-S micro, Xevo TQ-S cronos, SYNAPT XS, and SELECT SERIES Cyclic IMS mass spectrometers
  • Replaced as required
  • Does not include a Sample Cone
Catalog No. 50-239-1218 Supplier Waters Corp Supplier No. 700010291
This replacement Ion Block Sub Assembly for Stepwave mass spectrometers is a mechanical component designed to facilitate the entry of ions into the mass spectrometer prior to detection.
  • Mechanical type
  • Designed for MS systems
  • Facilitates ion entry into the mass spectrometer prior to detection
  • Compatible with Xevo TQ-XS, Xevo TQ-S micro, Xevo TQ-S cronos, SYNAPT XS, and SELECT SERIES Cyclic IMS mass spectrometers
  • Replaced as required
  • Does not include a Sample Cone