Learn More
Waters Corp WATERS CORPORATION Ion Block Support Assembly, PEEK, Non-Stepwave Mass Spectrometers
Catalog No. 50792953 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement PEEK Ion Block Support Assembly is designed to connect the Ion Block Assembly to various mass spectrometers.
  • Designed for replacement in mass spectrometers.
  • Connects the ion block assembly to non-stepwave mass spectrometers.
  • Compatible with SYNAPT G2, Xevo G2, Xevo QTof, Xevo TQ, Xevo TQD, and SQD2 mass spectrometers.
Catalog No. 50-792-953 Supplier Waters Corp Supplier No. 700004201
This replacement PEEK Ion Block Support Assembly is designed to connect the Ion Block Assembly to various mass spectrometers.
  • Designed for replacement in mass spectrometers.
  • Connects the ion block assembly to non-stepwave mass spectrometers.
  • Compatible with SYNAPT G2, Xevo G2, Xevo QTof, Xevo TQ, Xevo TQD, and SQD2 mass spectrometers.