Learn More
Waters Corp Ion Block Support Assembly, PEEK, for Stepwave Mass Spectrometers
Catalog No. 502391581 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Ion Block Support Assembly is designed for Stepwave mass spectrometers. Made from PEEK (Polyether ether ketone), it facilitates the connection of the Ion Block Assembly to specific mass spectrometers.
  • Designed for Stepwave mass spectrometers
  • Made from PEEK (Polyether ether ketone)
  • Facilitates connection of Ion Block Assembly
  • Compatible with Xevo TQ-S micro and Xevo TQ-S cronos mass spectrometers
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Type: Mechanical
  • System type: MS
  • Replacement schedule: As required
Catalog No. 50-239-1581 Supplier Waters Corp Supplier No. 700010294
This replacement Ion Block Support Assembly is designed for Stepwave mass spectrometers. Made from PEEK (Polyether ether ketone), it facilitates the connection of the Ion Block Assembly to specific mass spectrometers.
  • Designed for Stepwave mass spectrometers
  • Made from PEEK (Polyether ether ketone)
  • Facilitates connection of Ion Block Assembly
  • Compatible with Xevo TQ-S micro and Xevo TQ-S cronos mass spectrometers
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Type: Mechanical
  • System type: MS
  • Replacement schedule: As required