Learn More
Waters Corp Waters Corporation Ion Block Support Assembly, Replacement Part, Mechanical, MS
Catalog No. 50466647 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement ion block support assembly designed for Waters mass spectrometers. It serves as a crucial mechanical component for various MS systems.
  • Spare parts kit
  • One unit per package
  • Mechanical type
  • For MS system type
  • Replaced as required
  • Compatible with Synapt G2-S, Synapt G2-Si, Vion, Xevo G2-S, Xevo G2-XS, Xevo TQ-S, and Xevo TQ-XS mass spectrometers.
Catalog No. 50-466-647 Supplier Waters Corp Supplier No. 700005352
This is a replacement ion block support assembly designed for Waters mass spectrometers. It serves as a crucial mechanical component for various MS systems.
  • Spare parts kit
  • One unit per package
  • Mechanical type
  • For MS system type
  • Replaced as required
  • Compatible with Synapt G2-S, Synapt G2-Si, Vion, Xevo G2-S, Xevo G2-XS, Xevo TQ-S, and Xevo TQ-XS mass spectrometers.