Learn More
Waters Corp Ion Block Support Assembly, PEEK, Ceramic
Catalog No. 50792898 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Ion Block Support Assembly is a crucial component used in mass spectrometry systems, particularly ion mobility mass spectrometers. It facilitates the proper assembly and functioning of the ion block, ensuring precise operation and reliable performance in analytical applications.
  • Features a PEEK terminal block.
  • Includes a ceramic heater mounting block.
  • Designed for precise fitting of cover seals, cone gas O-rings, and isolation valve O-rings.
  • Critical for maintaining system integrity in ion mobility mass spectrometry.
  • Requires careful handling during assembly to prevent damage to delicate components.
Catalog No. 50-792-898 Supplier Waters Corp Supplier No. 700003662
The Ion Block Support Assembly is a crucial component used in mass spectrometry systems, particularly ion mobility mass spectrometers. It facilitates the proper assembly and functioning of the ion block, ensuring precise operation and reliable performance in analytical applications.
  • Features a PEEK terminal block.
  • Includes a ceramic heater mounting block.
  • Designed for precise fitting of cover seals, cone gas O-rings, and isolation valve O-rings.
  • Critical for maintaining system integrity in ion mobility mass spectrometry.
  • Requires careful handling during assembly to prevent damage to delicate components.