Learn More
Waters Corp Ion Chamber, CI, MS System, Replacement Part
Catalog No. 502391958 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Ion Chamber is designed for Chemical Ionization (CI) applications within an MS System. It functions as a critical component, used specifically on the Xevo TQ-GC system.
  • Categorized as a replacement parts kit
  • Mechanical type
  • Used in MS (Mass Spectrometry) systems
  • Replaced as required
Catalog No. 50-239-1958 Supplier Waters Corp Supplier No. 700012014
This replacement Ion Chamber is designed for Chemical Ionization (CI) applications within an MS System. It functions as a critical component, used specifically on the Xevo TQ-GC system.
  • Categorized as a replacement parts kit
  • Mechanical type
  • Used in MS (Mass Spectrometry) systems
  • Replaced as required