Learn More
Apexbio Technology LLC DiscoveryProbe™ Ion Channel Compound Library, 250uL/well(10 mM solution), 96 Well Racks With Screw Cap
SDP
Catalog No. 501905600
Encompass_Preferred
A unique collection of 199 ion channel-related compounds for high throughput screening (HTS) and high content screening (HCS). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5600 Supplier Apexbio Technology LLC Supplier No. L1030.25
Add to Cart
Add to Cart
Request a Quote
A unique collection of 199 ion channel-related compounds for high throughput screening (HTS) and high content screening (HCS). Other sizes are available. Please inqury us for quote.