Learn More
Enzo Life Sciences SCREEN-WELL Ion Channel ligand library (1 Library)
SDP
Catalog No. NC1682232
Encompass_Preferred
The SCREEN-WELL Ion Channel Ligand Library contains 70 ion channel blockers and openers for use in characterizing and identifying ion channels in individual cells or tissue. Application: HTS. Use/Stability: Stable for at least one year from the date of receipt when stored at -80°C. Long Term Storage: -80°C
Catalog No. NC1682232 Supplier Enzo Life Sciences Supplier No. BML28050100
The SCREEN-WELL Ion Channel Ligand Library contains 70 ion channel blockers and openers for use in characterizing and identifying ion channels in individual cells or tissue. Application: HTS. Use/Stability: Stable for at least one year from the date of receipt when stored at -80°C. Long Term Storage: -80°C