Learn More
Cole-Parmer Environmental Express Ion Exchange Columns, 2.5 mL with Frit and Cap; 50/Pk
Catalog No. NC1832117 Shop All Cole-Parmer Products
Encompass
The clarified polypropylene columns have a built-in polyethylene frit with nominal 30 m particle retention. The unique design sets the frit slightly above the bottom, creating a shoulder that increases surface area for more efficient resin separations. Order funnels separately for ease of fillfunnels attach directly to column entrance.
Catalog No. NC1832117 Supplier Cole-Parmer Supplier No. 3520042
The clarified polypropylene columns have a built-in polyethylene frit with nominal 30 m particle retention. The unique design sets the frit slightly above the bottom, creating a shoulder that increases surface area for more efficient resin separations. Order funnels separately for ease of fillfunnels attach directly to column entrance.