Learn More
Ace Glass, Inc. ION EXCHANGE ASSY C ION EXCHANGE ASSY C
SDP
Catalog No. 50972541
Encompass
Ion-Exchange assembly, designed for metal recovery using Ion-Exchange resins such as Amborane. Borosilicate glass column has a #25 Ace-Thred at both ends. Polypropylene end fittings form a leak-tight seal with FETFE O-Rings. Fittings have 1/4" NPT female thread for connecting 3/8" O.D. flexible vinyl tubing from pump and reservoir using a 1/4" to 3/8" tubing connector. Normal flow is in upward direction. Assembly consists column, pump, tubing, screen supports, O-ring(s), ring stand, clamp, screen supports, connector, and elbow tubes.
Catalog No. 50-972-541 Supplier Ace Glass, Inc. Supplier No. 5881502
Ion-Exchange assembly, designed for metal recovery using Ion-Exchange resins such as Amborane. Borosilicate glass column has a #25 Ace-Thred at both ends. Polypropylene end fittings form a leak-tight seal with FETFE O-Rings. Fittings have 1/4" NPT female thread for connecting 3/8" O.D. flexible vinyl tubing from pump and reservoir using a 1/4" to 3/8" tubing connector. Normal flow is in upward direction. Assembly consists column, pump, tubing, screen supports, O-ring(s), ring stand, clamp, screen supports, connector, and elbow tubes.