Learn More
Solstice Ion Exchange SPE Columns, Strong Cation Exchange (Silica Based) Benzenesulfonic Acd, 500 mg/6 mL
Catalog No. 6003088 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
30
Catalog No. Quantity
600-30-88 30
1 options
Catalog No. 600-30-88 Supplier Solstice Supplier No. 9092
Add to Cart
Add to Cart
Medical-grade polypropylene with PE frits
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 30