Learn More
Solstice Ion Exchange SPE Columns, Strong Cation Exchange (Silica Based) Benzenesulfonic Acid, 200 mg/3 mL
Catalog No. 6003083 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
600-30-83 50
1 options
Catalog No. 600-30-83 Supplier Solstice Supplier No. 9091
Add to Cart
Add to Cart
Medical-grade polypropylene with PE frits
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50