Learn More
VAISALA INC ION EXCHANGED WATER HMK15 60ML
Catalog No. NC3446032
Encompass_Preferred
Ion exchanged water for preparation of saturated salt solutions. 60ml bottle, sufficient for 4 chambers.
Catalog No. NC3446032 Supplier VAISALA INC Supplier No. 19767HM
Ion exchanged water for preparation of saturated salt solutions. 60ml bottle, sufficient for 4 chambers.