Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion exchanger Amberlyst™ 15, H+ form, EMD-Millipore

Catalog No. M1156350500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1156350500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1156350500 Supplier MilliporeSigma Supplier No. 1.15635.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strongly acid cation exchangers

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ion exchanger Amberlyst™ IR-120
Density 0.6g/cm3 (690kg/m3 Bulk Density)
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.