Learn More
Waters Corp Waters Corporation Ion Exclusion Column, 7 μm, 7.8 mm, 150 mm
Catalog No. 50784972 Shop All Waters Corporation Products
Encompass_Preferred
IC-Pak Ion Exclusion columns analyze weak acid anions and short-chain organic acids. They feature a durable bed for high-sensitivity analyses, using nominal 7 μm spherical particles of fully sulfonated resin for precise inorganic ion separation in complex matrices.
  • Durable column bed for repetitive, high-sensitivity analyses.
  • Nominal 7 μm spherical particles; fully sulfonated resin.
  • High precision and sensitivity for inorganic ion separation.
  • Withstands up to 2000 psi.
  • Ion exclusion phase ensures reliable reproducibility.
  • HPLC technology for superior results.
  • Manufactured to stringent requirements.
  • Functional for analytical and preparative scales.
  • Expands lab capabilities for weak acid anion separation.
Catalog No. 50-784-972 Supplier Waters Corp Supplier No. WAT010295
IC-Pak Ion Exclusion columns analyze weak acid anions and short-chain organic acids. They feature a durable bed for high-sensitivity analyses, using nominal 7 μm spherical particles of fully sulfonated resin for precise inorganic ion separation in complex matrices.
  • Durable column bed for repetitive, high-sensitivity analyses.
  • Nominal 7 μm spherical particles; fully sulfonated resin.
  • High precision and sensitivity for inorganic ion separation.
  • Withstands up to 2000 psi.
  • Ion exclusion phase ensures reliable reproducibility.
  • HPLC technology for superior results.
  • Manufactured to stringent requirements.
  • Functional for analytical and preparative scales.
  • Expands lab capabilities for weak acid anion separation.