Learn More
Waters Corp IC-Pak Ion-Exclusion Column, 7 µm, 7.8 mm x 300 mm, 1/pk
Catalog No. 50819144 Shop All Waters Corporation Products
Encompass_Preferred
IC-Pak Ion Exclusion columns are used for the analysis of weak acid anions such as fluoride and short chain organic acids from formate to butyrate.
Catalog No. 50-819-144 Supplier Waters Corp Supplier No. WAT010290
IC-Pak Ion Exclusion columns are used for the analysis of weak acid anions such as fluoride and short chain organic acids from formate to butyrate.