Learn More
Agilent Technologies Ion Injector, 180 mm, 0.6 mm ID, Dielectric, 1 ea
Catalog No. NC1976261
Encompass
The ion injector serves as a critical entry point for LC/MS systems, facilitating the transition of samples into the vacuum region while accelerating ions toward the focusing optics. By utilizing an opposing electrical charge, the injector draws ions through the nitrogen drying gas and into the sampling path. This dielectric model is designed with a 0.6 mm inner diameter and a length of 180 mm to ensure precise ion delivery and compatibility with various Agilent LC/MS instrument platforms.
  • Internal diameter of 0.6 mm
  • Length of 180 mm
  • Dielectric construction
  • Optimized for efficient ion transport and acceleration
  • Compatible with a wide range of Agilent 6100, 6200, 6400, and 6500 series LC/MS systems
Catalog No. NC1976261 Supplier Agilent Technologies Supplier No. G760460000
The ion injector serves as a critical entry point for LC/MS systems, facilitating the transition of samples into the vacuum region while accelerating ions toward the focusing optics. By utilizing an opposing electrical charge, the injector draws ions through the nitrogen drying gas and into the sampling path. This dielectric model is designed with a 0.6 mm inner diameter and a length of 180 mm to ensure precise ion delivery and compatibility with various Agilent LC/MS instrument platforms.
  • Internal diameter of 0.6 mm
  • Length of 180 mm
  • Dielectric construction
  • Optimized for efficient ion transport and acceleration
  • Compatible with a wide range of Agilent 6100, 6200, 6400, and 6500 series LC/MS systems