Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Ion S5™ Calibration Standard

Catalog No. A27988
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27988 8 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27988 Supplier Ion Torrent™ Supplier No. A27988
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Ion S5™ Calibration Standard is a small panel consisting of known sequence content with comprehensive and uniform representation of long homopolymers up to 10-mers.

The Ion S5™ Calibration Standard is a small panel consisting of known sequence content with comprehensive and uniform representation of long homopolymers up to 10-mers. The Ion S5 Calibration Standard can be used in conjunction with the Ion 520™ and Ion 530™ workflows for de novo research sequencing applications to allow for greater accuracy of base-calling.

Note:
This kit is only compatible with the Ion S5™ and Ion S5™ XL Sequencing Systems.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5°C to -30°C
For Use With (Application) Sequencing
No. of Reactions 8
Product Line GeneArt
Product Type Calibration Standard
Quantity 8 Reactions
Shipping Condition Dry Ice
Workflow Step Template Preparation
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.