Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Ion Library TaqMan™ Quantitation Kit

Catalog No. 4468802
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 reactions, 2 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-688-02 250 reactions, 2 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-688-02 Supplier Applied Biosystems™ Supplier No. 4468802
  • $2,110.00
    $1,856.65 / Each of 1

    Save $253.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

An easy-to-use, flexible solution for the accurate quantitation of library input for the Ion Torrent semiconductor sequencing workflow

  • Precision: The TaqMan™ Assay provides specificity and accuracy in detecting properly adapted, amplifiable templates over non-probe-based quantitation methods
  • Sensitivity: The protocol supports quantitation of femtomolar amounts of fragment libraries
  • Simplicity: The validated, ready-to-use qPCR standard requires only a simple serial dilution to determine the optimal concentration of library to use for templated bead preparation
  • Versatility: The highly robust qPCR mix accommodates a wide range of cycling conditions and reaction volumes, and combines highly sensitive detection with a broad quantification range
  • Efficiency: qPCR-based methods use less material for quantification
TRUSTED_SUSTAINABILITY

Specifications

Calibration Type Sequencing - Library Quantitation
Content And Storage Store components at -20° C
Format Kit
Excitation Visible
Emission Visible
For Use With (Equipment) Ion PGM™ System and Ion S5 Plus
For Use With (Application) Sequencing
No. of Reactions 250
Product Line Ion Torrent
Product Type Quantitation Kit
Quantity 250 reactions, 2 Tubes
Sample Volume 2.5 ml
Shipping Condition Wet Ice
Workflow Step Library Quantitation
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.